Assay Details
| Gene Symbol: SIRPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018556.3 | NP_061026.2 | 4 - 5 | 1139 | 147 | |
| XM_005260749.3 | XP_005260806.1 | 3 - 4 | 857 | 147 | ||
| XM_011529286.2 | XP_011527588.1 | 4 - 5 | 1186 | 147 | ||
| GenBank mRNA | AB042624.1 | - | 4 - 5 | 1155 | 147 | |
| AK292745.1 | - | 4 - 5 | 1139 | 147 | ||
| BC064532.1 | - | 4 - 5 | 1100 | 5588038 | 147 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000555.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20008057.hg.1 | ENST00000303415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008057.hg.1 | ENST00000381580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008057.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008057.hg.1 | TC20000555.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008165.hg.1 | ENST00000340424.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170926.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170926.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170926.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170926.hg.1 | TC2000009971.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000170926.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170926.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170926.hg.1 | ENST00000381580.5 | Ensembl |

SIRPG
Genomic Map