Assay Details
| Gene Symbol: CCDC144NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC068597.1 | - | 3 - 4 | 689 | 4837813 | 87 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007279.hg.1 | ENST00000562547.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007279.hg.1 | ENST00000562547.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007279.hg.1 | ENST00000618784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007279.hg.1 | ENST00000618784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007279.hg.1 | ENST00000618784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007279.hg.1 | ENST00000618784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007279.hg.1 | ENST00000618784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008769.hg.1 | ENST00000416563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008769.hg.1 | ENST00000416563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179073.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179073.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179073.hg.1 | TC1700010068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700179073.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179073.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007279.hg.1 | ENST00000562547.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007279.hg.1 | ENST00000562547.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007279.hg.1 | ENST00000618784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007279.hg.1 | ENST00000618784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007279.hg.1 | ENST00000618784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007279.hg.1 | ENST00000618784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007279.hg.1 | ENST00000618784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179075.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179075.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179075.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179075.hg.1 | TC1700010068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700179075.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179075.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179075.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179076.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179076.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179076.hg.1 | TC1700010068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700179076.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179076.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008769.hg.1 | ENST00000416563.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008769.hg.1 | ENST00000416563.1 | Ensembl |

CCDC144NL
Genomic Map