Assay Details
| Gene Symbol: RCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002901.2 | NP_002892.1 | 3 - 4 | 898 | 153 | |
| GenBank mRNA | AK094360.1 | - | 2 - 3 | 988 | 153 | |
| AK129791.1 | - | 3 - 4 | 925 | 153 | ||
| AY423728.1 | - | 1 - 2 | 134 | 153 | ||
| BC010120.2 | - | 3 - 4 | 767 | 3836263 | 153 | |
| D42073.1 | - | 3 - 4 | 684 | 153 | ||
| FJ224346.1 | - | 3 - 4 | 684 | 153 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000304.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000528630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000528630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000528630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000528630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000532721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000532721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000532721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003888.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003888.hg.1 | ENST00000532942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003888.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003888.hg.1 | TC11000304.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199990.hg.1 | TC1100012981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000527337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000528630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000528630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000528630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000528630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000531345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000532474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000532721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000532721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000532721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012981.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199987.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199987.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199987.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199987.hg.1 | ENST00000054950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199987.hg.1 | ENST00000533898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100199987.hg.1 | NM_002901.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100199987.hg.1 | TC1100012981.hg.1 | Clariom D TC |

RCN1
Genomic Map