| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008663.hg.1 |
ENST00000217270.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008663.hg.1 |
ENST00000546004.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008663.hg.1 |
NM_144773.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008663.hg.1 |
TC20000594.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008287.hg.1 |
ENST00000217270.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008287.hg.1 |
ENST00000217270.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008287.hg.1 |
NM_144773.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008287.hg.1 |
NM_144773.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008662.hg.1 |
ENST00000217270.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008662.hg.1 |
ENST00000546004.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008662.hg.1 |
NM_144773.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008662.hg.1 |
TC20000594.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000158398.hg.1 |
ENST00000217270.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158398.hg.1 |
NM_144773.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158398.hg.1 |
TC2000008287.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000158398.hg.1 |
ENST00000217270.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158398.hg.1 |
NM_144773.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158399.hg.1 |
TC2000008287.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC2000008287.hg.1 |
ENST00000217270.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008287.hg.1 |
ENST00000217270.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008287.hg.1 |
NM_144773.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008287.hg.1 |
NM_144773.3 |
Refseq |