Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox D12 | HOXD12 | HOX4H | -2.2kb | Anti-sense | Chr. 2 - 176099802 - 176100760 on NCBI Build GRCh38 |
| Downstream | homeobox D11 | HOXD11 |
HOX4, HOX4F |
1000.0 | Anti-sense | Chr. 2 - 176107356 - 176109588 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map