Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HOXC cluster antisense RNA 2 | HOXC-AS2 | Anti-sense | Chr. 12 - 53995208 - 53996785 on NCBI Build GRCh38 | ||
| Downstream | microRNA 615 | MIR615 |
MIRN615, hsa-mir-615, mir-615 |
5.0kb | Sense | Chr. 12 - 54033950 - 54034045 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map