Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox D12 | HOXD12 | HOX4H | -5.0kb | Sense | Chr. 2 - 176099802 - 176100760 on NCBI Build GRCh38 |
| Downstream | homeobox D9 | HOXD9 |
HOX4, HOX4C, Hox-4.3, Hox-5.2 |
3.4kb | Sense | Chr. 2 - 176122685 - 176124917 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map