Assay Details
| Gene Symbol: HOXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006712477.2 | XP_006712540.1 | 1 - 2 | 65 | 81 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BQ722165.1 | - | 39 | Chr. - - [-] on Build | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HOXD cluster antisense RNA 2 | HOXD-AS2 | Anti-sense | Chr. 2 - 176134841 - 176137098 on NCBI Build GRCh38 | ||
| Downstream | HOXD antisense growth-associated long non-coding RNA | HAGLR |
HOXD-AS1, Mdgt |
3.5kb | Anti-sense | Chr. 2 - 176173189 - 176188958 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006712477.2 transcripts.
Set Membership: |
|

HOXD3
Genomic Map