Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HOXA11 antisense RNA | HOXA11-AS |
HOXA-AS5, HOXA11-AS1, HOXA11AS, HOXA11S, NCRNA00076 |
Anti-sense | Chr. 7 - 27185408 - 27189293 on NCBI Build GRCh38 | |
| Downstream | HOXA distal transcript antisense RNA | HOTTIP |
HOXA-AS6, HOXA13-AS1, NCRNA00213 |
7.0kb | Anti-sense | Chr. 7 - 27200421 - 27207259 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map