Assay Details
| Gene Symbol: PCDHGB8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001297.1 | 1 | 1153 | 140 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protocadherin gamma subfamily A, 1 |
PCDHGA1 | PCDH-GAMMA-A1 | Sense | Chr. 5 - 141330685 - 141512979 on NCBI Build GRCh38 | |
| Downstream |
protocadherin gamma subfamily A, 12 |
PCDHGA12 |
CDH21, FIB3, PCDH-GAMMA-A12 |
2.3kb | Sense | Chr. 5 - 141430591 - 141512979 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P177531 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P177531 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 37579 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 28644 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P177531 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P177531 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_19_P00317266 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327625 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327615 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327625 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327615 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04327619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24506261 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P177531 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P177531 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327687 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327554 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327570 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327567 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327551 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327589 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327627 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327550 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327579 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327632 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327653 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327548 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327561 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327659 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327674 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327586 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327672 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327644 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327664 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327577 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327619 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327587 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327681 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327615 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327634 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327629 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327679 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327617 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327658 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327666 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327591 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327685 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327600 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327662 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15747620 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327604 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327565 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327543 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15747617 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327677 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15747619 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327675 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327602 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327624 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327694 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327654 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327575 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327670 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327613 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327556 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327656 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15747622 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327622 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327608 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327611 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327606 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327691 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327560 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327620 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327541 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327648 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327633 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327559 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04327650 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 234029_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234029_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234029_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234029_at5 | - | - |
| Human Genome U133 Set | 234029_PM_at | - | - |
| Human Genome U133 Set | 234029_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028957 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028957 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1803296 | NR_001297.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1803296 | NR_001297.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1803296 | NR_001297.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1803296 | NR_001297.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1803296 | NR_001297.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1803296 | NR_001297.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_5198 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_5198 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1803296 | NR_001297.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1803296 | NR_001297.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1803296 | NR_001297.1 | human_transcriptome |

PCDHGB8P
Genomic Map