Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gonadotropin releasing hormone receptor 2 (pseudogene) | GNRHR2 | GnRH-II-R | Sense | Chr. 1 - 145919013 - 145925341 on NCBI Build GRCh38 | |
| Downstream | limb and CNS expressed 1 like | LIX1L | 11.7kb | Anti-sense | Chr. 1 - 145933423 - 145958038 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map