Assay Details
| Gene Symbol: SNORA66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002444.2 | 1 | 65 | 102 | ||
| GenBank mRNA | Y11160.1 | - | 1 | 65 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 69 member A | FAM69A | Anti-sense | Chr. 1 - 92832729 - 92961522 on NCBI Build GRCh38 | ||
| Downstream | metal response element binding transcription factor 2 | MTF2 |
M96, PCL2, TDRD19A, dJ976O13.2 |
238.4kb | Sense | Chr. 1 - 93079219 - 93139081 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000854.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3779 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3778 | - | - |
| SurePrint G3 Human GE Microarray G4851A | ERCC-00148_99 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409058 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409076 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00379365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409065 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409058 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409065 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409076 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00379365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20409061 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379362 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379336 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379374 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379324 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379322 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379355 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379354 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379367 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379345 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379344 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379340 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379373 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379342 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379379 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379330 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379333 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379370 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379365 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379359 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379346 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379375 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379353 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379327 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379361 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379383 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379337 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379364 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379368 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379381 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00379320 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002444_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7903018 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7903010 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000383953.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000384792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NR_000006.8 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009046.hg.1 | NR_002444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013601.hg.1 | ENST00000384792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013601.hg.1 | NR_002444.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013601.hg.1 | TC01000854.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100164510.hg.1 | ENST00000384792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164510.hg.1 | NR_002444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164510.hg.1 | ENST00000384792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100164510.hg.1 | NR_002444.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100164510.hg.1 | TC0100009046.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000315741.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000370321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000383953.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000384792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000461952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000470843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | ENST00000497519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NM_000969.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NR_000006.8 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009046.hg.1 | NR_002444.2 | Refseq |
| Human Genome U133 Plus 2.0 Array | 210035_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 210035_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 210035_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 210035_s_at | - | - |
| Human Genome U133 Set | 210035_PM_s_at | - | - |
| Human Genome U133 Set | 210035_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1689616 | Y11160.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1689616 | Y11160.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1689616 | Y11160.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1689616 | Y11160.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1689616 | Y11160.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1689616 | Y11160.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1850646 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_28289 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1689616 | Y11160.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1689616 | Y11160.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1689616 | Y11160.1 | human_transcriptome |

SNORA66
Genomic Map