Assay Details
| Gene Symbol: TRPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002720.2 | 5 - 6 | 685 | 100 | ||
| GenBank mRNA | X89067.1 | - | 5 - 6 | 668 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927708 | LOC101927708 | -63.3kb | Anti-sense | Chr. 11 - 3507980 - 3571987 on NCBI Build GRCh38 | |
| Downstream | ADP-ribosyltransferase 5 | ART5 | ARTC5 | 2.3kb | Anti-sense | Chr. 11 - 3638503 - 3642392 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

TRPC2
Genomic Map