| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012908.hg.1 |
ENST00000473864.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012908.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012908.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012908.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007871.hg.1 |
ENST00000438325.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034643.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03034643.hg.1 |
TC03001937.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0300012908.hg.1 |
ENST00000473864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012908.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012908.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012908.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300196462.hg.1 |
ENST00000473864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300196462.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300196462.hg.1 |
ENST00000473864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300196462.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300196462.hg.1 |
TC0300012908.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300196463.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300196463.hg.1 |
NR_003284.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300196463.hg.1 |
TC0300012908.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900007871.hg.1 |
ENST00000438325.2 |
Ensembl |