Assay Details
| Gene Symbol: SNORD105B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003688.1 | 1 | 43 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PPAN-P2RY11 readthrough | PPAN-P2RY11 | Sense | Chr. 19 - 10106223 - 10115389 on NCBI Build GRCh38 | ||
| Downstream | purinergic receptor P2Y11 | P2RY11 | P2Y11 | 1.7kb | Sense | Chr. 19 - 10111521 - 10115389 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000171.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4246 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31494098 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31494099 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31494092 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880343 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31494097 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880337 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880336 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880340 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880344 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880338 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880345 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880339 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880342 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880336 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31494097 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31494098 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880345 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880339 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31494097 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880343 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31494099 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880341 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880340 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880336 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880345 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880338 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31494098 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880339 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880342 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31494092 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880344 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10880337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880345 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880339 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31494097 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10880336 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31494098 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880401 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880356 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880392 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880314 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880366 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880330 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880310 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880406 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880393 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880403 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880395 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880343 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880341 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880336 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880371 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17107267 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880377 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880374 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880379 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880316 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880365 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880321 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880361 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880329 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880332 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17107278 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17107262 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880398 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880312 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880317 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880384 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880320 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880387 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880363 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880333 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880354 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880322 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880368 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17107251 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880382 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880359 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880308 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880390 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17107271 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880381 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880339 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17107265 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880357 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880386 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880345 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10880315 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900006971.hg.1 | ENST00000458770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006971.hg.1 | NR_003688.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002616.hg.1 | ENST00000458770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002616.hg.1 | NR_003688.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002616.hg.1 | TC19000171.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900149530.hg.1 | ENST00000458770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149530.hg.1 | NR_003688.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900149530.hg.1 | TC1900006971.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900149530.hg.1 | ENST00000458770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149530.hg.1 | NR_003688.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006971.hg.1 | ENST00000458770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006971.hg.1 | NR_003688.1 | Refseq |

SNORD105B
Genomic Map