Assay Details
| Gene Symbol: SNORD123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003689.1 | 1 | 45 | 78 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BF063745.1 | - | 1 | Chr. 5 - 9548704 - 9549177 [-] on Build GRCh38 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small nucleolar RNA host gene 18 | SNHG18 | Sense | Chr. 5 - 9546200 - 9550298 on NCBI Build GRCh38 | ||
| Downstream | taste 2 receptor member 1 | TAS2R1 |
T2R1, TRB7 |
80.1kb | Anti-sense | Chr. 5 - 9628997 - 9630351 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000067.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23923484 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23923468 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03757955 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23923485 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03757956 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03757963 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23923488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23923483 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03757954 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23923477 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23923475 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23923467 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03757947 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23923476 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03757958 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 327958 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3590673 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P639679 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4248 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P639671 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4247 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 16937 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 22057 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 10702 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P15087052 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3590673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923440 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757929 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923457 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923436 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923459 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757937 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923443 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757926 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923453 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923462 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757941 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923435 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923429 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757907 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923460 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757909 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757923 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757928 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757939 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757913 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923438 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757925 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923441 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757934 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757939 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923440 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923453 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923436 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757913 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923460 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923459 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757909 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923435 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757925 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757941 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923441 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757929 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757926 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757928 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757934 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923457 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757907 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923443 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923429 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757923 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923462 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03757937 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23923438 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P639671 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P639679 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P639679 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P639671 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15087052 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P156445 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757908 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757877 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757955 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757893 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757919 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757874 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757889 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757929 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757903 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15662488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757923 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757936 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757861 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757886 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757846 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757915 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757867 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757891 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757906 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757855 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757958 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757933 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757945 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757866 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15662490 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757925 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757942 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757904 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757857 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757881 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757939 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757947 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757951 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757852 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757911 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757899 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757921 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15662492 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757870 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15662477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757894 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757884 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757865 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757848 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03757897 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P15087052 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 227655_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000750.hg.1 | ENST00000459473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000750.hg.1 | NR_003689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000750.hg.1 | TC05000067.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500145948.hg.1 | ENST00000459473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145948.hg.1 | NR_003689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145948.hg.1 | ENST00000459473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145948.hg.1 | NR_003689.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145948.hg.1 | TC0500006712.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 227655_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 227655_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227655_at7 | - | - |
| Human Genome U95 Set | 44628_PM_at | - | - |
| Human Genome U95 Set | 44628_at | - | - |
| Human Genome U95 Set | 83706_PM_at | - | - |
| Human Genome U95 Set | 83706_at | - | - |
| Human Genome U133 Set | 227655_at | - | - |
| Human Genome U133 Set | 227655_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1909156 | AK095831.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3238972 | AK095831.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3238972 | NR_003689.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1909156 | AK095831.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3238972 | AK095831.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3238972 | NR_003689.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238972 | AK095831.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238972 | NR_003689.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238972 | AK095831.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238972 | NR_003689.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1909156 | AK095831.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1190438 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_72400 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1909156 | AK095831.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1909156 | AK095831.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1909156 | AK095831.1 | human_transcriptome |

SNORD123
Genomic Map