Assay Details
| Gene Symbol: SNORD111B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037420.1 | 1 | 45 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | component of oligomeric golgi complex 4 | COG4 |
CDG2J, COD1 |
-5.9kb | Anti-sense | Chr. 16 - 70480569 - 70523554 on NCBI Build GRCh38 |
| Downstream |
small nucleolar RNA, C/D box 111 |
SNORD111 | HBII-82 | 8.4kb | Sense | Chr. 16 - 70538005 - 70538098 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000580.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096102 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096086 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637459 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16910197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096082 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637468 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637450 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637456 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096104 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637457 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096089 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096094 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096103 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637455 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637460 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637466 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637453 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637446 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096091 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637466 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096091 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096103 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637453 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096094 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637459 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637460 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096082 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096089 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637450 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637456 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096104 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16910197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637468 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096086 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637457 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637455 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10096102 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30637446 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096065 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096103 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096075 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096106 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096082 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096068 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096069 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096095 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096071 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096100 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096091 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096066 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096085 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16910197 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096079 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096087 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096129 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16910204 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096101 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096109 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16910198 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096073 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096078 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096064 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096093 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16910200 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096088 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096107 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10096090 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16008516.hg.1 | ENST00000408587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008516.hg.1 | NR_037420.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008516.hg.1 | TC16000580.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008276.hg.1 | ENST00000408587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008276.hg.1 | NR_037420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162448.hg.1 | ENST00000408587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162448.hg.1 | NR_037420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600162448.hg.1 | TC1600008276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600162448.hg.1 | ENST00000408587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600162448.hg.1 | NR_037420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008276.hg.1 | ENST00000408587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008276.hg.1 | NR_037420.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3241848 | NR_037420.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3241848 | NR_003696.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3241848 | NR_037420.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3241848 | NR_003696.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3241848 | NR_037420.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3241848 | NR_003696.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3241848 | NR_037420.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3241848 | NR_003696.1 | human_transcriptome |

SNORD111B
Genomic Map