Assay Details
| Gene Symbol: SNAR-G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_004383.1 | 1 | 66 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chorionic gonadotropin beta subunit 1 | CGB1 | -86.0 | Anti-sense | Chr. 19 - 49035569 - 49036934 on NCBI Build GRCh38 | |
| Downstream | chorionic gonadotropin beta subunit 5 | CGB5 |
CGB, HCG, hCGB |
6.7kb | Sense | Chr. 19 - 49043845 - 49045311 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000722.hg.1
Set Membership: |
|

SNAR-G1
Genomic Map