Assay Details
| Gene Symbol: H2BFXP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003238.2 | 3 - 4 | 624 | 120 | ||
| GenBank mRNA | AI493191.1 | - | 1 - 2 | 120 | ||
| Gene Symbol: LOC100101478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130172.1 | 3 - 4 | 624 | 120 | ||
| GenBank mRNA | AY730279.1 | - | 3 - 4 | 624 | 120 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thymosin beta 15B | TMSB15B | Tbeta15b | -11.0kb | Sense | Chr. X - 103962627 - 103965990 on NCBI Build GRCh38 |
| Downstream | solute carrier family 25 member 53 | SLC25A53 | MCART6 | 37.3kb | Sense | Chr. X - 104099214 - 104157027 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000506.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17383660 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17383461 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 754798 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 751116 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 317563 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 175308 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 260590 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 131981 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3402551 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P85591 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 36099 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 44957 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 17316 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P85591 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3402551 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3402551 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P85591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383461 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383416 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589042 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383415 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589049 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383415 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383416 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589042 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11859158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32589049 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17383461 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P85591 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17383415 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11859156 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11859159 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11859153 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17383414 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11859150 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11859152 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8169079 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8169078 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8169077 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8169073 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 230664_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 233963_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010441.hg.1 | ENST00000441761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010441.hg.1 | ENST00000441761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010441.hg.1 | ENST00000441761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006775.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006775.hg.1 | TC0X000506.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006776.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006776.hg.1 | TC0X000506.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000417637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000417637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000417637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000598335.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000598335.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | ENST00000598335.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006774.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006774.hg.1 | TC0X000506.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006777.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006777.hg.1 | TC0X000506.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00154816.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154816.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154816.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154816.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154816.hg.1 | TC0X00008023.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010441.hg.1 | ENST00000441761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010441.hg.1 | ENST00000441761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010441.hg.1 | ENST00000441761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00154817.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154817.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154817.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154817.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154817.hg.1 | TC0X00008023.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00154810.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154810.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154810.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154810.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154810.hg.1 | TC0X00008023.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00154814.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154814.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154814.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154814.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154814.hg.1 | TC0X00008023.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00154812.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154812.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154812.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154812.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154812.hg.1 | TC0X00008023.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000355016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000417637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000417637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000417637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000598335.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000598335.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | ENST00000598335.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008026.hg.1 | NM_001164416.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154809.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154809.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154809.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154809.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154809.hg.1 | TC0X00008023.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00154818.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154818.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154818.hg.1 | NR_003238.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154818.hg.1 | NR_130172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00154818.hg.1 | TC0X00008023.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 233963_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 233963_at | - | - |
| Human Genome U133 Plus 2.0 Array | 230664_at | - | - |
| Human Genome U133 Plus 2.0 Array | 230664_PM_at | - | - |
| Human Genome U95 Set | 64216_at | - | - |
| Human Genome U95 Set | 64216_PM_at | - | - |
| Human Genome U133 Set | 233963_PM_at | - | - |
| Human Genome U133 Set | 230664_at | - | - |
| Human Genome U133 Set | 230664_PM_at | - | - |
| Human Genome U133 Set | 233963_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041351 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040237 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041351 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040237 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2234730 | NR_003238.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2234730 | AY730279.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2234730 | NR_003238.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2234730 | AY730279.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2234730 | NR_003238.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2234730 | AY730279.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2234730 | NR_003238.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2234730 | AY730279.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2234730 | NR_003238.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2234730 | AY730279.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2234730 | NR_003238.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2234730 | AY730279.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2234730 | NR_003238.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2234730 | AY730279.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2234730 | NR_003238.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2234730 | AY730279.1 | human_transcriptome |

H2BFXP
Genomic Map