| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07022171.hg.1 |
ENST00000457166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07022171.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07022171.hg.1 |
TC07001466.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07022170.hg.1 |
ENST00000457166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07022170.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07022170.hg.1 |
TC07001466.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07022169.hg.1 |
ENST00000457166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07022169.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07022169.hg.1 |
TC07001466.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700180933.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180933.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180933.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700180933.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180933.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180933.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700180933.hg.1 |
TC0700011374.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700180934.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180934.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180934.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700180934.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180934.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180934.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700180934.hg.1 |
TC0700011374.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700180932.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180932.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700180932.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180932.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700180932.hg.1 |
TC0700011374.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000434721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011374.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700180937.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180937.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700180937.hg.1 |
ENST00000457166.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700180937.hg.1 |
NR_003934.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700180937.hg.1 |
TC0700011374.hg.1 |
Clariom D TC |