Assay Details
| Gene Symbol: GSTM2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002932.2 | 1 | 378 | 114 | ||
| GenBank mRNA | AY532924.1 | - | 1 | 345 | 114 | |
| AY532925.1 | - | 1 | 345 | 114 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosome production factor 2 homolog | RPF2 |
BXDC1, bA397G5.4 |
-18.2kb | Anti-sense | Chr. 6 - 110982017 - 111028263 on NCBI Build GRCh38 |
| Downstream | solute carrier family 16 member 10 | SLC16A10 |
MCT10, PRO0813, TAT1 |
39.9kb | Anti-sense | Chr. 6 - 111087503 - 111227125 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002014.hg.1
Set Membership: |
|

GSTM2P1
Genomic Map