Assay Details
| Gene Symbol: ACKR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122951.2 | NP_001116423.1 | 1 | 907 | 67 | |
| NM_002036.3 | NP_002027.2 | 2 | 1673 | 67 | ||
| GenBank mRNA | AF030521.1 | - | 1 | 732 | 67 | |
| AK291593.1 | - | 2 | 1673 | 67 | ||
| AK309521.1 | - | 1 | 1176 | 67 | ||
| AY167991.1 | - | 1 | 726 | 67 | ||
| BC095496.1 | - | 1 | 729 | 67 | ||
| BM671230.1 | - | 1 | 67 | |||
| BX537430.1 | - | 3 | 1570 | 67 | ||
| CA435450.1 | - | 1 | 67 | |||
| HE572751.1 | - | 2 | 726 | 67 | ||
| JX081310.1 | - | 1 | 732 | 67 | ||
| U01839.1 | - | 1 | 907 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ACKR1
Genomic Map