Assay Details
| Gene Symbol: KIR2DL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015868.2 | NP_056952.2 | 3 | 248 | 161 | |
| GenBank mRNA | AF135566.1 | - | 1 | 148 | 161 | |
| AF135567.1 | - | 1 | 148 | 161 | ||
| AF285435.1 | - | 2 | 171 | 161 | ||
| AY789057.1 | - | 3 | 211 | 161 | ||
| BC032422.1 | - | 3 | 224 | 5220339 | 161 | |
| BC050730.1 | - | 3 | 242 | 5219336 | 161 | |
| BC110613.1 | - | 3 | 225 | 40031619 | 161 | |
| FN395076.1 | - | 3 | 211 | 161 | ||
| FN552550.1 | - | 3 | 193 | 161 | ||
| L41268.1 | - | 3 | 248 | 161 | ||
| L76662.1 | - | 3 | 222 | 161 | ||
| L76663.1 | - | 3 | 222 | 161 | ||
| U24074.1 | - | 3 | 232 | 161 | ||
| U50546.1 | - | 3 | 222 | 161 | ||
| U96190.1 | - | 1 | 148 | 161 | ||
| Gene Symbol: KIR2DL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014219.2 | NP_055034.2 | 3 | 240 | 161 | |
| GenBank mRNA | AF285434.1 | - | 2 | 171 | 161 | |
| AY366242.1 | - | 3 | 240 | 161 | ||
| KP260924.1 | - | 3 | 207 | 161 | ||
| L76669.1 | - | 3 | 222 | 161 | ||
| U24075.1 | - | 3 | 225 | 161 | ||
| Gene Symbol: KIR2DS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291695.1 | NP_001278624.1 | 3 | 246 | 161 | |
| NM_001291701.1 | NP_001278630.1 | 3 | 246 | 161 | ||
| NM_012312.4 | NP_036444.1 | 3 | 246 | 161 | ||
| GenBank mRNA | AF135565.1 | - | 1 | 148 | 161 | |
| AF285438.1 | - | 2 | 171 | 161 | ||
| AJ002102.1 | - | 3 | 211 | 161 | ||
| AJ002103.1 | - | 3 | 211 | 161 | ||
| AJ002104.1 | - | 3 | 211 | 161 | ||
| AJ002105.1 | - | 3 | 211 | 161 | ||
| AM409094.1 | - | 3 | 212 | 161 | ||
| AY366243.1 | - | 3 | 221 | 161 | ||
| AY366244.1 | - | 3 | 210 | 161 | ||
| BC108917.1 | - | 3 | 213 | 40017299 | 161 | |
| BC119631.1 | - | 3 | 230 | 40115568 | 161 | |
| BC119632.1 | - | 3 | 230 | 40115570 | 161 | |
| BC125146.1 | - | 2 | 195 | 161 | ||
| BC126856.1 | - | 3 | 213 | 40017301 | 161 | |
| FR750994.1 | - | 3 | 246 | 161 | ||
| L41347.1 | - | 3 | 230 | 161 | ||
| L76667.1 | - | 3 | 222 | 161 | ||
| U24079.1 | - | 3 | 209 | 161 | ||
| X89893.1 | - | 3 | 211 | 161 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

KIR2DL3
Genomic Map