Assay Details
Gene Symbol: ST7-AS2 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109980.1 | 4 - 5 | 1050 | 140 | |||
GenBank mRNA | AF400041.1 | - | 3 - 4 | 845 | 140 | ||
AF400042.1 | - | LIT2001 ST7OT2 |
4 - 5 | 1071 | 140 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 6132 | MIR6132 | hsa-mir-6132 | -51.8kb | Anti-sense | Chr. 7 - 117020211 - 117020319 on NCBI Build GRCh38 |
Downstream | ST7 overlapping transcript 3 | ST7-OT3 |
NCRNA00026, ST7, ST7OT3 |
109.5kb | Anti-sense | Chr. 7 - 117182681 - 117209937 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700189057.hg.1 | TC0700012363.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
