Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927708 | LOC101927708 | -56.8kb | Sense | Chr. 11 - 3507980 - 3571987 on NCBI Build GRCh38 | |
| Downstream | ADP-ribosyltransferase 5 | ART5 | ARTC5 | 4.1kb | Sense | Chr. 11 - 3638503 - 3642392 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map