Assay Details
| Gene Symbol: SNRPN | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF400497.1 | - | LIT2205 IC-SNURF-SNRPN 3 prime noncoding transcripts |
13 - 14 | 1881 | 107 | |
| AF400498.1 | - | LIT2221 IC-SNURF-SNRPN 3 prime noncoding transcripts |
11 - 12 | 973 | 107 | ||
| AY362866.1 | - | 5 - 6 | 565 | 107 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 115-23 |
SNORD115-23 | HBII-52-23 | Sense | Chr. 15 - 25211796 - 25211877 on NCBI Build GRCh38 | |
| Downstream |
small nucleolar RNA, C/D box 115-24 |
SNORD115-24 | HBII-52-24 | 1.5kb | Sense | Chr. 15 - 25213659 - 25213729 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNRPN
Genomic Map