Assay Details
| Gene Symbol: DLEU1 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ412047.1 | - | LIT1893 BCMS |
1 - 2 | 219 | 73 | |
| AJ412048.1 | - | LIT1892 BCMS |
1 - 2 | 183 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 4 | ST13P4 |
FAM10A4, FAM10A4P |
-136.5kb | Sense | Chr. 13 - 50172018 - 50173616 on NCBI Build GRCh38 |
| Downstream | DLEU1 antisense RNA 1 | DLEU1-AS1 | LINC01308 | 130.6kb | Anti-sense | Chr. 13 - 50520933 - 50527449 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

DLEU1
Genomic Map