Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger MYM-type containing 3 | ZMYM3 |
DXS6673E, MYM, XFIM, ZNF198L2, ZNF261 |
Anti-sense | Chr. X - 71239624 - 71255261 on NCBI Build GRCh38 | |
| Downstream |
non-POU domain containing, octamer-binding |
NONO |
MRXS34, NMT55, NRB54, P54, P54NRB, PPP1R114 |
31.4kb | Sense | Chr. X - 71283192 - 71301168 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map