Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506302 | LOC100506302 | -57.7kb | Sense | Chr. 7 - 155641916 - 155644406 on NCBI Build GRCh38 | |
| Downstream | sonic hedgehog | SHH |
HHG1, HLP3, HPE3, MCOPCB5, SMMCI, TPT, TPTPS |
99.6kb | Sense | Chr. 7 - 155799984 - 155812273 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map