Assay Details
| Gene Symbol: SNORD42B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_000013.1 | 1 | 38 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB34, member RAS oncogene family |
RAB34 |
RAB39, RAH |
-2.3kb | Anti-sense | Chr. 17 - 28714281 - 28718268 on NCBI Build GRCh38 |
| Downstream |
small nucleolar RNA, C/D box 4A |
SNORD4A |
RNU101A, Z17A, mgh18S-121 |
2.0kb | Sense | Chr. 17 - 28722582 - 28722653 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000326.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3663 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_19_P00813545 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10273446 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10273447 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30837731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30837728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10273446 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30837728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10273447 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30837731 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16966010 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16966023 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273436 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273483 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273486 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273416 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273495 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273491 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273460 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273434 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273485 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16966001 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273492 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273489 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273497 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273459 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16965994 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273418 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273480 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273446 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273438 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273469 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10273498 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16966012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_000013_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8005952 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8005951 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000458893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000459083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000459584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000496182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000496182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000496182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000496182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000496182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000580755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000580755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000580755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000580755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000580755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000582736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000582736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | ENST00000582736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | NR_000009.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | NR_000013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007383.hg.1 | NR_000014.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004009.hg.1 | ENST00000458893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004009.hg.1 | NR_000013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004009.hg.1 | TC17000326.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700152585.hg.1 | ENST00000458893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152585.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152585.hg.1 | NR_000013.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152585.hg.1 | TC1700007383.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700152585.hg.1 | ENST00000458893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152585.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152585.hg.1 | NR_000013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000355731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394935.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000394938.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000422514.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000458893.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000459083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000459584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000472628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000496182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000496182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000496182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000496182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000496182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000578181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000580755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000580755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000580755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000580755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000580755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000582736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000582736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | ENST00000582736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | NM_000984.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | NR_000009.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | NR_000013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007383.hg.1 | NR_000014.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1810762 | NR_000013.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1810762 | NR_000013.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1810762 | NR_000013.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1810762 | NR_000013.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1810762 | NR_000013.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1810762 | NR_000013.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 2000019 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_1411 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1810762 | NR_000013.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1810762 | NR_000013.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1810762 | NR_000013.1 | human_transcriptome |

SNORD42B
Genomic Map