| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19010306.hg.1 |
ENST00000363660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19010306.hg.1 |
NR_001285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19010306.hg.1 |
TC19000736.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000363660.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000599561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008542.hg.1 |
NR_001285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900166113.hg.1 |
ENST00000363660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900166113.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900166113.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900166113.hg.1 |
NR_001285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900166113.hg.1 |
TC1900008542.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900166113.hg.1 |
ENST00000363660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900166113.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900166113.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900166113.hg.1 |
NR_001285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000270625.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000363660.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000594493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000596873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599167.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000599561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000600027.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000601306.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
ENST00000602252.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
NM_001015.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008542.hg.1 |
NR_001285.1 |
Refseq |