| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023763.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023763.hg.1 |
ENST00000527780.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023763.hg.1 |
ENST00000534096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023763.hg.1 |
ENST00000534789.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023763.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023763.hg.1 |
TC19001745.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023765.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023765.hg.1 |
ENST00000527780.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023765.hg.1 |
ENST00000534096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023765.hg.1 |
ENST00000534789.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023765.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023765.hg.1 |
TC19001745.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023764.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023764.hg.1 |
ENST00000527780.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023764.hg.1 |
ENST00000534096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023764.hg.1 |
ENST00000534789.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023764.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023764.hg.1 |
TC19001745.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
TC1900011195.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194014.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000531692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011195.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
TC1900011195.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194012.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
NR_002798.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
TC1900011195.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
ENST00000525179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
ENST00000527780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
ENST00000534096.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
ENST00000534789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
ENST00000562112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900194013.hg.1 |
NR_002798.1 |
Refseq |