Assay Details
| Gene Symbol: PDLIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256425.1 | NP_001243354.1 | 3 - 4 | 432 | 119 | |
| GenBank mRNA | BC026029.2 | - | 2 - 3 | 294 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hematopoietic prostaglandin D synthase | HPGDS |
GSTS, GSTS1, GSTS1-1, PGD2, PGDS |
-180.8kb | Anti-sense | Chr. 4 - 94298556 - 94342876 on NCBI Build GRCh38 |
| Downstream | BMPR1B antisense RNA 1 (head to head) | BMPR1B-AS1 | 170.3kb | Anti-sense | Chr. 4 - 94743668 - 94757533 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001256425.1 transcripts.
Set Membership: |
|

PDLIM5
Genomic Map