Assay Details
| Gene Symbol: SNX29P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002939.3 | 1 - 2 | 108 | 99 | ||
| GenBank mRNA | BX648280.1 | - | 3 - 4 | 780 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | translation initiation factor IF-2-like | LOC107984833 | -11.3kb | Sense | Chr. 16 - 29289365 - 29290955 on NCBI Build GRCh38 | |
| Downstream | nuclear pore complex interacting protein family member B11 | NPIPB11 | NPIP | 68.7kb | Anti-sense | Chr. 16 - 29381318 - 29403802 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000317.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16004507.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004507.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004507.hg.1 | TC16000317.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004506.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004506.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004506.hg.1 | TC16000317.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600153433.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153433.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153433.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153433.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600153433.hg.1 | TC1600007392.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600153433.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153433.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153433.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153433.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600153432.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153432.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153432.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600153432.hg.1 | TC1600007392.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600153432.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153432.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153432.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |

SNX29P2
Genomic Map