| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8054053 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8054048 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8054047 |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02008843.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02008843.hg.1 |
TC02000578.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200158900.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158900.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158900.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158900.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158900.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158900.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158900.hg.1 |
TC0200008493.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200158899.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158899.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158899.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158899.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158899.hg.1 |
TC0200008493.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200158901.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158901.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158901.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158901.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158901.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158901.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158901.hg.1 |
TC0200008493.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000427863.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
ENST00000607780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008493.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158897.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158897.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158897.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158897.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158897.hg.1 |
ENST00000443258.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158897.hg.1 |
ENST00000467292.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158897.hg.1 |
ENST00000483000.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158897.hg.1 |
NR_003698.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158897.hg.1 |
TC0200008493.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
228198_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
228199_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
222055_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
228198_PM_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
222055_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
228199_PM_at |
- |
- |
| Human Genome U133A 2.0 Array |
222055_at |
- |
- |
| Human Genome U133A 2.0 Array |
222055_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228199_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
228198_s_at7 |
- |
- |
| Human Genome U95 Set |
47085_at |
- |
- |
| Human Genome U95 Set |
47085_PM_at |
- |
- |
| Human Genome U95 Set |
78702_PM_at |
- |
- |
| Human Genome U95 Set |
78702_at |
- |
- |
| Human Genome U133 Set |
222055_at |
- |
- |
| Human Genome U133 Set |
222055_PM_at |
- |
- |
| Human Genome U133 Set |
228199_PM_at |
- |
- |
| Human Genome U133 Set |
228198_s_at |
- |
- |
| Human Genome U133 Set |
228199_at |
- |
- |
| Human Genome U133 Set |
228198_PM_s_at |
- |
- |