| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000476992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000476992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000476992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000476992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000476992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013332.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000781.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000781.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000781.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000781.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000781.hg.1 |
TC07003318.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196279.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196279.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196279.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196279.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196279.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196279.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196279.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196279.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196279.hg.1 |
TC0700013332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700196278.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196278.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196278.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196278.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196278.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196278.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196278.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196278.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196278.hg.1 |
TC0700013332.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000353796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396904.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000396912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000476992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000476992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000476992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000476992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000476992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
ENST00000498308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204456.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_001204513.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NM_021163.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013332.hg.1 |
NR_015343.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013333.hg.1 |
ENST00000407184.5 |
Ensembl |