Assay Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BM285394.1 | - | 4 - 5 | 1080 | 78 | |
| BQ635335.1 | - | 4 - 5 | 1147 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NSFL1 cofactor | NSFL1C |
P47, UBX1, UBXD10, UBXN2C, dJ776F14.1 |
-6.1kb | Sense | Chr. 20 - 1442162 - 1467720 on NCBI Build GRCh38 |
| Downstream | signal regulatory protein delta | SIRPD |
PTPNS1L2, dJ576H24.4 |
56.8kb | Sense | Chr. 20 - 1534251 - 1557697 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |

SIRPB2
Genomic Map