Assay Details
| Gene Symbol: KRTAP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_021489.1 | 1 - 2 | 323 | 116 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927562 | LOC101927562 | -16.3kb | Anti-sense | Chr. 11 - 1554607 - 1556457 on NCBI Build GRCh38 | |
| Downstream | keratin associated protein 5-3 | KRTAP5-3 |
KRTAP5-9, KRTAP5.3 |
8.4kb | Anti-sense | Chr. 11 - 1607565 - 1608463 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000045.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11000945.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000945.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000945.hg.1 | TC11000045.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000947.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000947.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000947.hg.1 | TC11000045.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000534077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000534077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013066.hg.1 | ENST00000398531.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013066.hg.1 | ENST00000398531.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013066.hg.1 | ENST00000398531.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013066.hg.1 | ENST00000398531.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013066.hg.1 | ENST00000398531.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013066.hg.1 | NM_001012710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013066.hg.1 | NM_001012710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013066.hg.1 | NM_001012710.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013066.hg.1 | NM_001012710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | ENST00000534077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | ENST00000534077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145549.hg.1 | TC1100006522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013066.hg.1 | ENST00000398531.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013066.hg.1 | ENST00000398531.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013066.hg.1 | ENST00000398531.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013066.hg.1 | ENST00000398531.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013066.hg.1 | ENST00000398531.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013066.hg.1 | NM_001012710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013066.hg.1 | NM_001012710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013066.hg.1 | NM_001012710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013066.hg.1 | NM_001012710.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000534077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000534077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145544.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145544.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145544.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145544.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145544.hg.1 | TC1100006522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100145550.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145550.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145550.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145550.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145550.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145550.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145550.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145550.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145550.hg.1 | TC1100006522.hg.1 | Clariom D TC |

KRTAP5-AS1
Genomic Map