Assay Details
| Gene Symbol: SNORD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003029.2 | 1 | 66 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromodomain helicase DNA binding protein 8 | CHD8 |
AUTS18, HELSNF1 |
Sense | Chr. 14 - 21385194 - 21437298 on NCBI Build GRCh38 | |
| Downstream |
small nucleolar RNA, C/D box 8 |
SNORD8 |
RNU6C, mgU6-53 |
5.0kb | Sense | Chr. 14 - 21397293 - 21397401 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000918.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16684029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16684035 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158409 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158401 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158405 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158408 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16684024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16684032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158399 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158389 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16684027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158389 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158399 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158408 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158409 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16684032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16684029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16684024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158405 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29615204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16684027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09158401 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16684035 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158332 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158435 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158339 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158422 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158406 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158430 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158409 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158347 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158437 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158334 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158456 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16684031 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158396 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158455 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158395 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158343 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158440 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16684026 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16684025 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158424 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158341 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158399 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16684015 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158408 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158431 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158447 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16684032 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158393 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16684012 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158452 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158420 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158426 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158413 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158459 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158410 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158340 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158345 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158453 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158337 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158432 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158390 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158415 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158349 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16684036 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158350 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158402 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158404 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158450 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16684011 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158443 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16684017 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09158352 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Genome U219 Array | 11763925_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11763925_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11763924_at | - | - |
| Affymetrix Human Genome U219 Array | 11763924_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008632.hg.1 | ENST00000362566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008632.hg.1 | NR_003029.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14010042.hg.1 | ENST00000362566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14010042.hg.1 | NR_003029.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14010042.hg.1 | TC14000918.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1400008632.hg.1 | ENST00000362566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008632.hg.1 | NR_003029.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162412.hg.1 | ENST00000362566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162412.hg.1 | NR_003029.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162412.hg.1 | ENST00000362566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400162412.hg.1 | NR_003029.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400162412.hg.1 | TC1400008632.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3244449 | NR_003029.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3244449 | NR_003029.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3244449 | NR_003029.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3244449 | NR_003029.2 | human_transcriptome |

SNORD9
Genomic Map