Assay Details
| Gene Symbol: NMRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_023352.1 | 2 - 3 | 275 | 125 | ||
| GenBank mRNA | AK097144.1 | - | 2 - 3 | 248 | 125 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | carnosine N-methyltransferase 1 | CARNMT1 |
C9orf41, UPF0586 |
-49.9kb | Sense | Chr. 9 - 74980794 - 75028456 on NCBI Build GRCh38 |
| Downstream | osteoclast stimulating factor 1 | OSTF1 |
OSF, SH3P2, bA235O14.1 |
5.3kb | Anti-sense | Chr. 9 - 75088480 - 75147201 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001220.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK097144.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015650.hg.1 | ENST00000376794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015650.hg.1 | ENST00000376811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015650.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015650.hg.1 | TC09001220.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900171481.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900171481.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900171481.hg.1 | ENST00000376811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900171481.hg.1 | NR_023352.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900171481.hg.1 | TC0900010445.hg.1 | Clariom D TC |

NMRK1
Genomic Map