Assay Details
| Gene Symbol: MRS2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024072.2 | 1 | 2148 | 86 | ||
| GenBank mRNA | AY333753.1 | - | 2147 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TBC1 domain family member 15 | TBC1D15 | RAB7-GAP | Anti-sense | Chr. 12 - 71839707 - 71926849 on NCBI Build GRCh38 | |
| Downstream | tryptophan hydroxylase 2 | TPH2 |
ADHD7, NTPH |
87.9kb | Anti-sense | Chr. 12 - 71938846 - 72032441 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001729.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3265444 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4456 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4457 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 17875 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3372281 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3265444 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3265444 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441278 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876643 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441278 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876635 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876646 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08441279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28876641 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441249 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441225 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441222 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441228 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441217 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441285 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441248 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441266 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441282 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441240 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441234 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441287 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441246 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441220 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441236 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441278 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441269 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441242 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441226 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441274 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441276 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441280 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441231 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441223 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08441244 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12022977.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12022977.hg.1 | TC12001729.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011262.hg.1 | ENST00000550293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011262.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011262.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011262.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187340.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187340.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187340.hg.1 | TC1200011262.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200187341.hg.1 | ENST00000550293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187341.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187341.hg.1 | ENST00000550293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187341.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187341.hg.1 | TC1200011262.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200187342.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187342.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187342.hg.1 | TC1200011262.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011262.hg.1 | ENST00000550293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011262.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011262.hg.1 | NR_024072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011262.hg.1 | NR_024072.2 | Refseq |
| Human Genome U95 Set | 59607_f_at | - | - |
| Human Genome U95 Set | 59607_PM_f_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047812 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047812 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3238254 | NR_024072.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3238254 | AY333753.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3238254 | NR_024072.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3238254 | AY333753.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238254 | NR_024072.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3238254 | AY333753.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238254 | NR_024072.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3238254 | AY333753.1 | human_transcriptome |

MRS2P2
Genomic Map