Assay Details
| Gene Symbol: ESRRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243515.1 | NP_001230444.1 | 3 - 4 | 525 | 84 | |
| NM_001243519.1 | NP_001230448.1 | 3 - 4 | 529 | 84 | ||
| XM_011509276.1 | XP_011507578.1 | 3 - 4 | 518 | 84 | ||
| XM_011509280.2 | XP_011507582.1 | 2 - 3 | 363 | 84 | ||
| GenBank mRNA | AK290945.1 | - | 3 - 4 | 506 | 84 | |
| BC008218.2 | - | 3 - 4 | 389 | 84 | ||
| DA722064.1 | - | 3 - 4 | 481 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | usherin | USH2A |
RP39, US2, USH2, dJ1111A8.1 |
-253.7kb | Sense | Chr. 1 - 215622894 - 216423396 on NCBI Build GRCh38 |
| Downstream | G-patch domain containing 2 | GPATCH2 |
CT110, GPATC2, PPP1R30 |
712.4kb | Sense | Chr. 1 - 217426992 - 217631312 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003822.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290945.1,BC008218.2,NM_001243515.1,NM_001243519.1,XM_011509276.1,XM_011509280.2 transcripts.
Set Membership: |
|

ESRRG
Genomic Map