| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007981.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007981.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007981.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007981.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007981.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007981.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007981.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007981.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004600.hg.1 |
ENST00000440586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004600.hg.1 |
ENST00000451980.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004600.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004600.hg.1 |
TC21000407.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004599.hg.1 |
ENST00000440586.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004599.hg.1 |
ENST00000451980.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004599.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004599.hg.1 |
TC21000407.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2100007981.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007981.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007981.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007981.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007981.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007981.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007981.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007981.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153798.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153798.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153798.hg.1 |
TC2100007981.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100153798.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153798.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153797.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153797.hg.1 |
NR_024102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153797.hg.1 |
TC2100007981.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100153797.hg.1 |
ENST00000451980.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153797.hg.1 |
NR_024102.1 |
Refseq |