| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030823.hg.1 |
ENST00000328902.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030823.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030823.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030823.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030823.hg.1 |
TC07002021.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030824.hg.1 |
ENST00000328902.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030824.hg.1 |
ENST00000392862.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030824.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030824.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030824.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030824.hg.1 |
TC07002021.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030827.hg.1 |
ENST00000328902.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030827.hg.1 |
ENST00000392862.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030827.hg.1 |
ENST00000477013.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030827.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030827.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030827.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030827.hg.1 |
TC07002021.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030825.hg.1 |
ENST00000328902.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030825.hg.1 |
ENST00000392862.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030825.hg.1 |
ENST00000477013.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030825.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030825.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030825.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030825.hg.1 |
TC07002021.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030830.hg.1 |
ENST00000328902.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030830.hg.1 |
ENST00000392862.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030830.hg.1 |
ENST00000493969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030830.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030830.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030830.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030830.hg.1 |
TC07002021.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193905.hg.1 |
TC0700013043.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013043.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193902.hg.1 |
TC0700013043.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193906.hg.1 |
TC0700013043.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
ENST00000328902.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
ENST00000493969.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
ENST00000618759.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
NM_001244071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
NM_001244072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
NM_024711.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193904.hg.1 |
TC0700013043.hg.1 |
Clariom D TC |