Assay Details
| Gene Symbol: GHRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302821.1 | NP_001289750.1 | 2 - 3 | 319 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ghrelin opposite strand/antisense RNA | GHRLOS |
GHRL-AS1, GHRLAS, NCRNA00068 |
Anti-sense | Chr. 3 - 10280952 - 10293449 on NCBI Build GRCh38 | |
| Downstream |
SEC13 homolog, nuclear pore and COPII coat complex component |
SEC13 |
D3S1231E, SEC13L1, SEC13R, npp-20 |
9.5kb | Sense | Chr. 3 - 10300929 - 10321188 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302821.1 transcripts.
Set Membership: |
|

GHRL
Genomic Map