| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
53885 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
280623 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
146893 |
AK074221.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
146893 |
AK125404.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
146893 |
BC006168.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
146893 |
BC028726.2 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
146893 |
D42047.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
146893 |
NM_015141.2 |
refseq_mrna_human_NM |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_32_P89730 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_32_P198810 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
16757 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
4625 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
60727 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
4447 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_32_P89730 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_32_P198810 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_32_P89730 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_32_P198810 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28628602 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08195314 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08195322 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08195318 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28628603 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28628606 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08195321 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08195322 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28628603 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08195321 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28628606 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08195318 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P28628602 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08195314 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P19459794 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K G4112F |
A_32_P89730 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K G4112F |
A_32_P198810 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19459794 |
- |
- |
| Agilent Human 1B Microarray G4111A |
A_32_P198810 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195297 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195304 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195322 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195316 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195298 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195288 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195296 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16535115 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195292 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195320 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195302 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195293 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195313 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195315 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195306 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08195300 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19459794 |
- |
- |