Assay Details
| Gene Symbol: LINC00486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027098.1 | 2 - 3 | 294 | 78 | ||
| GenBank mRNA | AK095182.1 | - | 2 - 3 | 294 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetratricopeptide repeat domain 27 | TTC27 | -20.0kb | Sense | Chr. 2 - 32628020 - 32821051 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100271832 | LOC100271832 | 28.5kb | Sense | Chr. 2 - 32927127 - 32946136 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000217.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003208.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003208.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003208.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003208.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003208.hg.1 | TC02000217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003206.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003206.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003206.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003206.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003206.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003206.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003206.hg.1 | TC02000217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003207.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003207.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003207.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003207.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003207.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003207.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003207.hg.1 | TC02000217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003199.hg.1 | TC02000217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003197.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003197.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003197.hg.1 | TC02000217.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149715.hg.1 | TC0200007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149707.hg.1 | TC0200007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149716.hg.1 | TC0200007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200149697.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149697.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149697.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149697.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149697.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149697.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149697.hg.1 | TC0200007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149717.hg.1 | TC0200007199.hg.1 | Clariom D TC |

LINC00486
Genomic Map