Assay Details
| Gene Symbol: LOC100130654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK095243.1 | - | 1 | 948 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | keratin 42 pseudogene | KRT42P | -8.9kb | Sense | Chr. 17 - 41626327 - 41640199 on NCBI Build GRCh38 | |
| Downstream | eukaryotic translation initiation factor 1 | EIF1 |
A121, EIF-1, EIF1A, ISO1, SUI1 |
37.4kb | Anti-sense | Chr. 17 - 41688875 - 41691646 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323633 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323609 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895892 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323632 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323600 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895940 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323620 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895883 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895914 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323629 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323643 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323638 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323648 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16982584 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323615 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895938 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323602 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323637 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323597 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323625 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323608 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323628 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323613 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16982592 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895909 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323630 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895930 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323631 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323601 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895936 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323636 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323610 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323616 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323650 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895889 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895941 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323646 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323635 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30895915 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323634 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323619 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10323606 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3253460 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P340010 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 37895 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4323 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3253460 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P340010 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P340010 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12647030 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P20865342 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P751111 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1562017_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562017_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562017_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562017_at6 | - | - |

LOC100130654
Genomic Map