Assay Details
| Gene Symbol: LOC646976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096082.1 | - | 1 | 153 | 84 | |
| Gene Symbol: RALGPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006711410.3 | XP_006711473.1 | 1 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4424 | MIR4424 | -46.5kb | Anti-sense | Chr. 1 - 178677749 - 178677834 on NCBI Build GRCh38 | |
| Downstream | angiopoietin like 1 | ANGPTL1 |
ANG3, ANGPT3, ARP1, AngY, UNQ162, dJ595C2.2 |
123.3kb | Sense | Chr. 1 - 178849535 - 178871353 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006711410.3 transcripts.
Set Membership: |
|

LOC646976
Genomic Map