Assay Details
| Gene Symbol: LOC283112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096599.1 | - | 1 - 2 | 61 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 6 member 5 | SLC6A5 |
GLYT-2, GLYT2, HKPX3, NET1 |
-6.9kb | Anti-sense | Chr. 11 - 20599400 - 20659285 on NCBI Build GRCh38 |
| Downstream | anoctamin 5 | ANO5 |
GDD1, LGMD2L, TMEM16E |
1.5mb | Anti-sense | Chr. 11 - 22192485 - 22283367 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC283112
Genomic Map